Learn More
LabScientific 25-place premium microscope slide box. Blue. Storage and transport of standard slides 1x3". Durable cork lining. Numbered slots match line inventory sheet.
Catalog No. NC1025494
Encompass
25-place premium microscope slide box. (Blue) Durable ABS plastic box securely stores slides in grooved, labeled slots and closes tightly. The molded feet enable secure stacking for easy transport and storage. Both the lining and the slots are numbered to correspond to the slide inventory sheet. Dimensions: L x W x H: 141 x 88 x 35 mm
Catalog No. NC1025494 Supplier LabScientific Supplier No. SB25B
25-place premium microscope slide box. (Blue) Durable ABS plastic box securely stores slides in grooved, labeled slots and closes tightly. The molded feet enable secure stacking for easy transport and storage. Both the lining and the slots are numbered to correspond to the slide inventory sheet. Dimensions: L x W x H: 141 x 88 x 35 mm