Learn More
Ted Pella Inc Tall Slide Box, 25 slides (3 x 2" slides)
SDP
Catalog No. NC1149191
Encompass
The Tall Slide Box made of sturdy ABS material is ideal for storage and transport of tall 3 x 2" (75 x 50mm) microscope slides. Available in a 100-slide and a 25-slide capacity box with grooved slots to prevent slide to slide contact. The base contains location numbers for convenient inventory, a cork liner in the 100-slide box and a foam liner in the 25-slide box. Lid interior contains a numbered index sheet for cataloging and identification. The deep design of the Tall Slide Box provides safe storage for the taller, non-standard microscope slides.
Catalog No. NC1149191 Supplier Ted Pella Inc Supplier No. 2195
The Tall Slide Box made of sturdy ABS material is ideal for storage and transport of tall 3 x 2" (75 x 50mm) microscope slides. Available in a 100-slide and a 25-slide capacity box with grooved slots to prevent slide to slide contact. The base contains location numbers for convenient inventory, a cork liner in the 100-slide box and a foam liner in the 25-slide box. Lid interior contains a numbered index sheet for cataloging and identification. The deep design of the Tall Slide Box provides safe storage for the taller, non-standard microscope slides.