Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 25(R,S)-Ruscogenin | 874485-32-2 | 5 MG
SDP

Catalog No. 5000419479
Encompass_Preferred

25(R,S)-Ruscogenin is a PI3K/Akt/mTOR signaling pathway inhibitor and TLR4 signaling inhibitor. It has been shown to suppress hepatocellular carcinoma (HCC) metastasis by reducing the expression of MMP-2, MMP-9, uPA, VEGF, and HIF-1α via regulating the PI3K/Akt/mTOR signaling pathway. Additionally, Ruscogenin alleviates LPS-induced pulmonary endothelial cell apoptosis by suppressing TLR4 signaling.

  • Molecular weight is 430.62.
  • Formula is C27H42O4.
  • Appears as a white to off-white solid.
  • Classified as a steroid.
  • Initial sources include plants (Liliaceae Liriope platyphylla Wang et Tang) and microorganisms (Bacillus megaterium CGMCC 1.1741).

Catalog No. 50-004-19479 Supplier Medchemexpress LLC Supplier No. HYN51365MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

25(R,S)-Ruscogenin is a PI3K/Akt/mTOR signaling pathway inhibitor and TLR4 signaling inhibitor. It has been shown to suppress hepatocellular carcinoma (HCC) metastasis by reducing the expression of MMP-2, MMP-9, uPA, VEGF, and HIF-1α via regulating the PI3K/Akt/mTOR signaling pathway. Additionally, Ruscogenin alleviates LPS-induced pulmonary endothelial cell apoptosis by suppressing TLR4 signaling.

  • Molecular weight is 430.62.
  • Formula is C27H42O4.
  • Appears as a white to off-white solid.
  • Classified as a steroid.
  • Initial sources include plants (Liliaceae Liriope platyphylla Wang et Tang) and microorganisms (Bacillus megaterium CGMCC 1.1741).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.