Learn More
Cayman Chemical 25(R)-27-hydroxy Cholesterol, Purity: ≥95%, CAS: 20380-11-4, 1 mg
SDP
Catalog No. 502973328
Encompass_Preferred
The more predominant-occurring (~80%) stereoisomer of the endogenous oxysterol 27-hydroxy cholesterol
Catalog No. 50-297-3328 Supplier Cayman Chemical Supplier No. 147901 MG
The more predominant-occurring (~80%) stereoisomer of the endogenous oxysterol 27-hydroxy cholesterol