Learn More
Cayman Chemical 25 R27hydroxy CholesTRol 10mg
SDP
Catalog No. 502969792
Encompass_Preferred
The more predominant-occurring (~80%) stereoisomer of the endogenous oxysterol 27-hydroxy cholesterol
Catalog No. 50-296-9792 Supplier Cayman Chemical Supplier No. 1479010 MG
The more predominant-occurring (~80%) stereoisomer of the endogenous oxysterol 27-hydroxy cholesterol