Learn More
Cayman Chemical 25(S)-27-hydroxy Cholesterol, Purity: ≥95%, CAS: 56845-83-1, 5 mg
SDP
Catalog No. 502969145
Encompass_Preferred
The less predominant-occurring (~20%) stereoisomer of the endogenous oxysterol 27-hydroxy cholesterol
Catalog No. 50-296-9145 Supplier Cayman Chemical Supplier No. 147915 MG
The less predominant-occurring (~20%) stereoisomer of the endogenous oxysterol 27-hydroxy cholesterol