Learn More
Cayman Chemical 25 S27hydroxy CholesTRol 10mg
SDP
Catalog No. 502968976
Encompass_Preferred
The less predominant-occurring (~20%) stereoisomer of the endogenous oxysterol 27-hydroxy cholesterol
Catalog No. 50-296-8976 Supplier Cayman Chemical Supplier No. 1479110 MG
The less predominant-occurring (~20%) stereoisomer of the endogenous oxysterol 27-hydroxy cholesterol