Learn More
Chemglass Life Sciences Silicone Sealing Ring, fits GL-25 Cap, fits tubing O.D. 7.5mm to 9.0mm
SDP
Catalog No. CG19707
Encompass_Preferred
Silicone Sealing Ring, fits GL-25 Cap, fits tubing O.D. 7.5mm to 9.0mm. Seal is made from silicone with a vulcanized PTFE washer having an aperture to fit the listed tubing size. For use with CG-195 cap.
Catalog No. CG19707 Supplier Chemglass Life Sciences Supplier No. CG19707
Silicone Sealing Ring, fits GL-25 Cap, fits tubing O.D. 7.5mm to 9.0mm. Seal is made from silicone with a vulcanized PTFE washer having an aperture to fit the listed tubing size. For use with CG-195 cap.