Learn More
Electron Microscopy Sciences 20-Slide Staining Dish Complete Set, 3¾" x 3" x 2¼" (H)
SDP
Catalog No. 50931042
Encompass_Preferred
These glass staining dishes are made of a heavy glass with snug fit lids. Lids are included. Dishes have a gasket seal. Includes one staining dish with lid and one handle rack. Accomodates 20 slides.
I.D.: 3¾" x 3" x 2¼" (H)
Dimensions: 4-1/4" x 3⅝" x 3" (H)
Catalog No. 50-931-042 Supplier Electron Microscopy Sciences Supplier No. 7142025
These glass staining dishes are made of a heavy glass with snug fit lids. Lids are included. Dishes have a gasket seal. Includes one staining dish with lid and one handle rack. Accomodates 20 slides.
I.D.: 3¾" x 3" x 2¼" (H)
Dimensions: 4-1/4" x 3⅝" x 3" (H)