Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions 25% SULFURIC ACID V/V 4L
SDP

Catalog No. NC0579150
Encompass_Preferred

Sulfuric Acid 25% V/v Solution (4l)

Catalog No. NC0579150 Supplier Aqua Solutions Supplier No. 90064L
May include imposed supplier surcharges.
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfuric Acid 25% V/v Solution (4l)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.