Learn More
Biolegend APC anti-human CD235a (Glycophorin A)
Supplier:  Biolegend 349113
Encompass_Preferred
APC anti-human CD235a (Glycophorin A) [HI264]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. NC1383028