Learn More
Chemglass Life Sciences Vacuum Trap, 25mm Body OD x 200mm Body Length, 10mm OD Side and Inner Tubes
SDP
Catalog No. CG451001
Encompass_Preferred
Vacuum Trap, 25mm Body OD x 200mm Body Length, 10mm OD Side and Inner Tubes. One piece trap used in the removal of undesired vapors in the vacuum system by condensation. 25mm Body OD x 200mm Body Length, 10mm OD Side and Inner Tubes.
Catalog No. CG451001 Supplier Chemglass Life Sciences Supplier No. CG451001
Vacuum Trap, 25mm Body OD x 200mm Body Length, 10mm OD Side and Inner Tubes. One piece trap used in the removal of undesired vapors in the vacuum system by condensation. 25mm Body OD x 200mm Body Length, 10mm OD Side and Inner Tubes.