Learn More
Accuflow Systems Saint-Gobain C-Flex Tubing, 0.250 in x 0.375 in, 350 ft
Catalog No. 502365815
Encompass_Preferred
C-Flex TPE tubing (Formulation 374) is a thermoplastic elastomer tubing supplied on 350 ft mini spools for high-purity bioprocessing and single-use fluid transfer. It is formulated for peristaltic pump compatibility, aseptic welding, and common sterilization processes.
  • Translucent thermoplastic elastomer construction.
  • Inner diameter 0.250 in, outer diameter 0.375 in, wall thickness 0.062-0.063 in.
  • Supplied as a 350 ft mini spool, single pack.
  • Suitable for peristaltic pump use and single-use assemblies.
  • Compatible with aseptic welding and sealing operations.
  • Sterilizable by autoclave (one-time 121°C/30 min) and by gamma or X-ray irradiation up to 50 kGy (one-time).
Catalog No. 50-236-5815 Supplier Accuflow Systems Supplier No. 3742502M
C-Flex TPE tubing (Formulation 374) is a thermoplastic elastomer tubing supplied on 350 ft mini spools for high-purity bioprocessing and single-use fluid transfer. It is formulated for peristaltic pump compatibility, aseptic welding, and common sterilization processes.
  • Translucent thermoplastic elastomer construction.
  • Inner diameter 0.250 in, outer diameter 0.375 in, wall thickness 0.062-0.063 in.
  • Supplied as a 350 ft mini spool, single pack.
  • Suitable for peristaltic pump use and single-use assemblies.
  • Compatible with aseptic welding and sealing operations.
  • Sterilizable by autoclave (one-time 121°C/30 min) and by gamma or X-ray irradiation up to 50 kGy (one-time).