Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 250 μL Automation Tips for Agilent™/Velocity 11 VPrep™ and Bravo, 96 Tip Format
GREENER_CHOICE

Catalog No. 14222881 Shop All Axygen Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Tip Style:
Standard
Features:
Non-pyrogenic
Non-pyrogenic, Bottom Label
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Tip Style Features
14-222-881 Non-sterile Standard Non-pyrogenic
14-222-884 Sterile Standard Non-pyrogenic
14-222-882 Non-sterile Standard Non-pyrogenic, Bottom Label
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-222-881 Supplier Axygen™ Supplier No. VT250R
Add to Cart
Add to Cart

For use with Agilent/Velocity11 automation systems

Tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.

  • Clear polypropylene
  • Maxymum Recovery™ improves accuracy and precision while reducing sample loss
  • Certified RNase-/DNase-free and nonpyrogenic
  • Nonpyrogenic
  • Available in nonsterile or sterile

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 250 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Certifications/Compliance ANSI
Clean Claims DNase, Rnase Free
For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls
Features Non-pyrogenic
Format Rack
Tip Style Standard
Packaging SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) Agilent/Velocity VPrep 8LT, Agilent/Velocity VPrep 96LT, Agilent Bravo 96LT
Material Polypropylene
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.