Learn More
Analytical Sales & Services 250µL GC Syringe for LEAP/Gerstel CTC PAL GC Autosamplers
Catalog No. 5000806410
Encompass
250µL GC Syringe 1725N for LEAP/Gerstel CTC PAL GC Autosamplers.
26 Gauge, 51mm Length, Poinst Style AS
Qty/Pkg: EACH
Catalog No. 50-008-06410 Supplier Analytical Sales & Services Supplier No. 42030784
250µL GC Syringe 1725N for LEAP/Gerstel CTC PAL GC Autosamplers.
26 Gauge, 51mm Length, Poinst Style AS
Qty/Pkg: EACH