Learn More
Labtech Inc 250ml evaporating flask
Catalog No. NC1125652
Encompass_Preferred
250ml evaporating flask, 29/32 NS, fits LabTech rotary evaporators
Catalog No. NC1125652 Supplier Labtech Inc Supplier No. LT501030250
250ml evaporating flask, 29/32 NS, fits LabTech rotary evaporators