Learn More
Ace Glass, Inc. 250mL complete gas washing bottle, porosity C
SDP
Supplier:  Ace Glass, Inc. 716316
Encompass_Preferred
250mL tall form gas washing bottle with "C" porosity fritted disc and 29/42 stopper.
Catalog No. NC1682749