Learn More
Cole-Parmer Pyrex Vista 70022-250 Graduated Glass Cylinder, 250 mL, to contain, 1/ea
Catalog No. NC9839395 Shop All Cole-Parmer Products
Encompass_Preferred
Manufactured of Class A borosilicate glass, these metric scale cylinders are enameled with blue graduations for easy reading. Cylinder capacity is in accordance wtih ASTM E-1272. 10mL size has a funnel top for easy filling.
This item is not returnable. View return policy
Catalog No. NC9839395 Supplier Cole-Parmer Supplier No. 3450454
Manufactured of Class A borosilicate glass, these metric scale cylinders are enameled with blue graduations for easy reading. Cylinder capacity is in accordance wtih ASTM E-1272. 10mL size has a funnel top for easy filling.