Learn More
Chemrus Inc 250 ML LIU FLASK 4/PACK
Catalog No. NC1444290
Encompass
LIU FLASK, FLAT BOTTOM, BARREL-SHAPE, HEAVY WALL, SINGLE NECK, 250ml, 24/40 taper outer joint, 4/pack
Catalog No. NC1444290 Supplier Chemrus Inc Supplier No. CR2401250
LIU FLASK, FLAT BOTTOM, BARREL-SHAPE, HEAVY WALL, SINGLE NECK, 250ml, 24/40 taper outer joint, 4/pack