Learn More
US Plastic Corporation 250 ML PP BEAKERS 50PK250 ML
SDP
Catalog No. NC0176916
Encompass
Disposable, low cost, unbreakable and very corrosive resistant. For use up to 250 deg. F. Autoclavable. Dripless lip and flat base. Graduated in ml and oz. Sold in package lots only. Translucent.
Catalog No. NC0176916 Supplier US Plastic Corporation Supplier No. 76056
Add to Cart
Add to Cart
Disposable, low cost, unbreakable and very corrosive resistant. For use up to 250 deg. F. Autoclavable. Dripless lip and flat base. Graduated in ml and oz. Sold in package lots only. Translucent.