Learn More
Perkin Elmer US LLC 250 mm, 4.6 mm I.D., 5 Quasar AQ LC Column
Catalog No. 502113014
Encompass_Preferred
Quasar AQ LC columns benefit from no phase collapse even in 100% aqueous mobile phase conditions and the polar end capping aids in retention of more polar compounds.
Catalog No. 50-211-3014 Supplier Perkin Elmer US LLC Supplier No. N9308840
Quasar AQ LC columns benefit from no phase collapse even in 100% aqueous mobile phase conditions and the polar end capping aids in retention of more polar compounds.