Learn More
NEST SCIENTIFIC INC 250 μl Robotic Tips for Beckman, Clear, Sterile, 50/cs
Catalog No. NC3650184
Encompass_Preferred
250uL automation tips for use with Beckman liquid handlers. Sterile and packed in 12X8 SBS/ANSI footprint racks. Lot tested in house on Beckman i7 to ensure proper fit and CV values. Molded in ISO 7 cleanroom and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. Produced in a cGMP (21 CFR part 820/821), ISO 13485 and ISO 9001 facility. 4800 tips per case.
Catalog No. NC3650184 Supplier NEST SCIENTIFIC INC Supplier No. 317511
250uL automation tips for use with Beckman liquid handlers. Sterile and packed in 12X8 SBS/ANSI footprint racks. Lot tested in house on Beckman i7 to ensure proper fit and CV values. Molded in ISO 7 cleanroom and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. Produced in a cGMP (21 CFR part 820/821), ISO 13485 and ISO 9001 facility. 4800 tips per case.