Learn More
Waters Corp Waters Corporation Syringe, 250 μL, High Pressure, Sample
Catalog No. NC9539624 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Corporation 250 μL Sample Syringe is a high-pressure syringe designed for use as a spare part kit, specifically for ACQUITY systems.
  • Product type: spare parts kits
  • System type: MS, UPLC
  • Replacement schedule: as required
Catalog No. NC9539624 Supplier Waters Corp Supplier No. 410001347
The Waters Corporation 250 μL Sample Syringe is a high-pressure syringe designed for use as a spare part kit, specifically for ACQUITY systems.
  • Product type: spare parts kits
  • System type: MS, UPLC
  • Replacement schedule: as required