Learn More
Perkin Elmer US LLC 250 uL Sampling Syringe
Supplier:  Perkin Elmer US LLC 09923270
Encompass_Preferred
The correct syringe is critical to ensure proper fit and operation within your instrument. Syringes must ensure a tight and secure fit in the injection port and withstand repeated injection cycles without bending or breaking. Syringes must deliver accurate and precise volumes with no dead volume or carryover between injections. Each syringe is individually inspected for accuracy and performance.
Catalog No. 50-211-7127