Learn More
ANALYTICAL SCIENTIFIC INSTRUMENTS US INC 250 µL Standard HPLC Static Mixer, Binary Tee Housing Assembly
SDP
Catalog No. NC1698608
Encompass
Maximum Operating Pressure: 6,000 PSI, Maximum Temperature Rating: 40 °C, Delay volume: 250 µL, Low Flow Series cartridges: 50 µL through 250 µL are interchangeable and fit in all Low Flow Series In-line, Binary tee and Ternary tee housings.
Catalog No. NC1698608 Supplier ANALYTICAL SCIENTIFIC INSTRUMENTS US INC Supplier No. 4120250
Maximum Operating Pressure: 6,000 PSI, Maximum Temperature Rating: 40 °C, Delay volume: 250 µL, Low Flow Series cartridges: 50 µL through 250 µL are interchangeable and fit in all Low Flow Series In-line, Binary tee and Ternary tee housings.