Learn More
Waters Corp Waters Syringe, 2500 μL, HPLC
Catalog No. 50819034 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This syringe is an optional component designed for use when an auxiliary loop is installed in the injector. It is specifically utilized for processing samples exceeding 200 μL.
  • Optional syringe used when the auxiliary loop is installed in the injector.
  • The auxiliary loop is used for samples greater than 200 μL.
Catalog No. 50-819-034 Supplier Waters Corp Supplier No. WAT077342
This syringe is an optional component designed for use when an auxiliary loop is installed in the injector. It is specifically utilized for processing samples exceeding 200 μL.
  • Optional syringe used when the auxiliary loop is installed in the injector.
  • The auxiliary loop is used for samples greater than 200 μL.