Learn More
Perkin Elmer US LLC 2500 Run Kit for Standard 75 mL, 40 Bar Digestion Vessels
Catalog No. 502111693
Encompass_Preferred
2500 Run Kit for Standard 75 mL (40 Bar) Digestion Vessels for Titan MPS 16 Position Microwave Sample Preparation Systems. This kit includes: 10 Pressure Seals (N3132002) 4 Rupture Discs (N3132001) 5 Lip Seal Rings of the Gas Containment Manifold (N3134000)
Catalog No. 50-211-1693 Supplier Perkin Elmer US LLC Supplier No. N3132000
2500 Run Kit for Standard 75 mL (40 Bar) Digestion Vessels for Titan MPS 16 Position Microwave Sample Preparation Systems. This kit includes: 10 Pressure Seals (N3132002) 4 Rupture Discs (N3132001) 5 Lip Seal Rings of the Gas Containment Manifold (N3134000)