Learn More
Perkin Elmer US LLC 2500 uL Sampling Syringe
Catalog No. 502117137
Encompass_Preferred
Syringes from PerkinElmer are individually inspected for accuracy and performance. Recommended autosampler syringes are available in 0.5, 5 and 50 µL capacities. For routine analyses, the metal plunger in barrel with PTFE-tipped seal is the standard syringe as shipped with each Clarus GC instrument. Alternative syringes to use are the metal plunger in barrel or the 0.53 mm on-column injection.
Catalog No. 50-211-7137 Supplier Perkin Elmer US LLC Supplier No. 09923219
Syringes from PerkinElmer are individually inspected for accuracy and performance. Recommended autosampler syringes are available in 0.5, 5 and 50 µL capacities. For routine analyses, the metal plunger in barrel with PTFE-tipped seal is the standard syringe as shipped with each Clarus GC instrument. Alternative syringes to use are the metal plunger in barrel or the 0.53 mm on-column injection.