Learn More
Chemglass Life Sciences Low Form Dewar Flask, Cylindrical, 2500mL Capacity, Fits 1000mL Round Bottom Flask (RBF) , 170mm ID X 135mm Inside Depth, 210mm OD X 185mm Height
SDP
Catalog No. CG159204
Encompass_Preferred
Low Form Dewar Flask, Cylindrical, 2500mL Capacity, Fits 1000mL RBF, 170mm ID X 135mm Inside Depth, 210mm OD X 185mm Height. Similar to CG-1590, but with a semi-flat bottom which increases coolant capacity and permits magnetic stirring of contents of vessel. With aluminum base that extends up to the top rim for added protection. Flask size shown is the maximum capacity flask that will fit inside the Dewar.
Catalog No. CG159204 Supplier Chemglass Life Sciences Supplier No. CG159204
Low Form Dewar Flask, Cylindrical, 2500mL Capacity, Fits 1000mL RBF, 170mm ID X 135mm Inside Depth, 210mm OD X 185mm Height. Similar to CG-1590, but with a semi-flat bottom which increases coolant capacity and permits magnetic stirring of contents of vessel. With aluminum base that extends up to the top rim for added protection. Flask size shown is the maximum capacity flask that will fit inside the Dewar.