Learn More
Chemglass Life Sciences Dish, Crystallizing, 2500mL, 190 x 100mm
SDP
Catalog No. 501215014
Encompass_Preferred
Dish, Crystallizing, 2500mL, 190 x 100mm. Tops are reinforced and fire polished to reduce chipping. Will withstand repeated sterilization.
Catalog No. 50-121-5014 Supplier Chemglass Life Sciences Supplier No. CG8276P190
Dish, Crystallizing, 2500mL, 190 x 100mm. Tops are reinforced and fire polished to reduce chipping. Will withstand repeated sterilization.