Learn More
Ace Glass, Inc. 2500mL epoxy coated reactor bottle hydrogenation/gas
SDP
Supplier:  Ace Glass, Inc. 747837
Encompass
2500mL Borosilicate glass bottle with tooled neck and epoxy coated to help prevent scratching. Bottles and stoppers sold separately. Working capacity is 1/2 actual capacity. Rated at 60 psi @ 20C.
Catalog No. 50-979-257