Learn More
US Plastic Corporation US Plastic Corp. Round Jar, 2500 mL, HDPE, UN Rated, Red Lid
SDP
Catalog No. NC3973767
Encompass
This round jar from US Plastic Corp. has a capacity of 2500 mL. Made from high-density polyethylene (HDPE), it is UN Rated and features a white jar with a red lid. It is suitable for various applications requiring secure storage and transport.
  • Capacity of 2500 mL
  • Constructed from high-density polyethylene (HDPE)
  • Features a white jar and red lid
  • UN Rated with rating 1H2/X3,3/S
  • Weighs 6.61 lb
Catalog No. NC3973767 Supplier US Plastic Corporation Supplier No. 56629
This round jar from US Plastic Corp. has a capacity of 2500 mL. Made from high-density polyethylene (HDPE), it is UN Rated and features a white jar with a red lid. It is suitable for various applications requiring secure storage and transport.
  • Capacity of 2500 mL
  • Constructed from high-density polyethylene (HDPE)
  • Features a white jar and red lid
  • UN Rated with rating 1H2/X3,3/S
  • Weighs 6.61 lb