Learn More
Ace Glass, Inc. 2500mL Buchner filter funnel with porosity B (70-100 micron) ACE fritted glass disc.
SDP
Catalog No. 50974294
Encompass_Preferred
2500mL Buchner filter funnel with porosity B (70-100 micron) ACE fritted glass disc.
Catalog No. 50-974-294 Supplier Ace Glass, Inc. Supplier No. 718684
2500mL Buchner filter funnel with porosity B (70-100 micron) ACE fritted glass disc.