Learn More
Ace Glass, Inc. 2500mL Buchner filter funnel with porosity E (4-8 micron) ACE fritted glass disc.
SDP
Catalog No. 50974296
Encompass_Preferred
2500mL Buchner filter funnel with porosity E (4-8 micron) ACE fritted glass disc.
Catalog No. 50-974-296 Supplier Ace Glass, Inc. Supplier No. 718690
2500mL Buchner filter funnel with porosity E (4-8 micron) ACE fritted glass disc.