Learn More
Waters Corp Smart Syringe, 2.5 mL, Headspace, Fixed Needle
Catalog No. 502391297 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 2500 μL Smart Syringe is a replacement component with a 2.5 mL capacity, designed for headspace applications and featuring a fixed needle. It is intended for the GC-PAL 3 system, with an annual replacement recommendation.
  • Capacity: 2.5 mL for various sample handling needs.
  • Designed for headspace applications, ensuring compatibility and optimal performance in relevant analytical processes.
  • Fixed needle design for robust and consistent analytical methods.
  • System compatibility with the GC-PAL 3 system.
  • Ensures reliability and adherence to manufacturing standards.
  • Annual replacement schedule provides guidance for maintenance and continued peak performance.
Catalog No. 50-239-1297 Supplier Waters Corp Supplier No. 700012539
The 2500 μL Smart Syringe is a replacement component with a 2.5 mL capacity, designed for headspace applications and featuring a fixed needle. It is intended for the GC-PAL 3 system, with an annual replacement recommendation.
  • Capacity: 2.5 mL for various sample handling needs.
  • Designed for headspace applications, ensuring compatibility and optimal performance in relevant analytical processes.
  • Fixed needle design for robust and consistent analytical methods.
  • System compatibility with the GC-PAL 3 system.
  • Ensures reliability and adherence to manufacturing standards.
  • Annual replacement schedule provides guidance for maintenance and continued peak performance.