Learn More
Refrigiwear Inc. 2509 REG BLK MED Dbl Layr Herringbne Grip Glove with finger tip coating
SDP
Supplier:  Refrigiwear Inc. 2509RBLKMED
Encompass_Preferred
When the cold weather requires you to “get a grip,” reach for these bad boys. Ready to tackle any task, these have a dipped coating on the thumb, index, & middle fingers, making them perfect for precision tasks like label peeling. The herringbone grip pattern on the palms adds an extra layer of confidence, ensuring a secure hold in any conditions. Plus, the silicone grip pattern performs better in the cold without cracking or getting slick. With their dual-layer construction & built-in liner, these winter work gloves provide both warmth & dexterity. Don't let the work at hand slip through your fingers—grab a pair of these grip-enhancing work gloves & conquer any job with ease. Sold by the dozen. Price shown is per pair. Dipped coating on thumb, index & middle fingers perfect for peeling labels Silicone herringbone grip pattern on palms Dual-layer outershell with built-in liner for extra warmth Sold by the dozen. Price shown is per pair.    
Catalog No. NC3867009