Learn More
Waters Corp Waters XBridge Premier Protein SEC Column, 250Å, 2.5 μm, 4.6 x 150 mm
Supplier:  Waters Corp 176005067
Encompass_Preferred
XBridge Premier Protein SEC Columns provide reliable protein size variant separations (10,000 to approx 650,000 Daltons) through the synergistic coupling of MaxPeak Premier High Performance Surfaces and BEH-PEO Particle technologies. This minimizes non-desired secondary interactions, reducing method development time and allowing for a “generic” method for many samples. One vial of mAb Size Variant Standard is included.
  • Reliable protein size variant separations (10,000 to approximately 650,000 daltons).
  • MaxPeak premier high performance surfaces and BEH-PEO particle technologies.
  • Minimized non-desired secondary ionic or hydrophobic interactions.
  • Allows for a “generic” or “platform-type” method for various samples.
  • Includes one vial of mAb size variant standard at no additional cost.
Catalog No. NC2172027