Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 250mg, Methyl 6-chloro-5-(4,4,5,5-tetramethyl-1,3,2- dioxaborolan-2-yl)pyridine-3-carboxylate, 97%, [1622217-33-7], M13128 297.542 C13H17BClNO4

Catalog No. 5038473
Encompass_Preferred

Compound: Methyl 6-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carboxylate

CAS#: 1622217-33-7

MDL#: not listed

Background: A chlorinated pyridine boronic ester bearing a methyl ester group, used as a Suzuki-Miyaura coupling partner in pharmaceutical research. The chlorine and ester groups both provide additional handles for sequential functionalization.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-384-73 Supplier Frontier Specialty Chemicals Supplier No. M13128250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Methyl 6-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carboxylate

CAS#: 1622217-33-7

MDL#: not listed

Background: A chlorinated pyridine boronic ester bearing a methyl ester group, used as a Suzuki-Miyaura coupling partner in pharmaceutical research. The chlorine and ester groups both provide additional handles for sequential functionalization.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.