Learn More
Ace Glass, Inc. 250mL 15mm equalizing add funnel
SDP
Catalog No. 50979249
Encompass_Preferred
250mL, 15mm equalizing addition funnel with graduations for use with ACE pressure reactors. Funnel is heavy walled and features a #15 Ace-Thred at the top, bottom drip tip, pressure equalizing arm, and 0-3mm threaded PTFE stopcock plug.
Catalog No. 50-979-249 Supplier Ace Glass, Inc. Supplier No. 729925
250mL, 15mm equalizing addition funnel with graduations for use with ACE pressure reactors. Funnel is heavy walled and features a #15 Ace-Thred at the top, bottom drip tip, pressure equalizing arm, and 0-3mm threaded PTFE stopcock plug.