Learn More
Ace Glass, Inc. Trap, rotary evaporator, self-washing, 250mL, 24/40 top, 14/20bottom
SDP
Catalog No. 50978861
Encompass_Preferred
250mL self-washing, rotary evaporator trap designed by Dr. William Boulanger. Design prevents blow-through and enables continuous addition with rotary evaporators. Trap has 24/40 top joint and 14/20 bottom joint.
Catalog No. 50-978-861 Supplier Ace Glass, Inc. Supplier No. 670315
250mL self-washing, rotary evaporator trap designed by Dr. William Boulanger. Design prevents blow-through and enables continuous addition with rotary evaporators. Trap has 24/40 top joint and 14/20 bottom joint.