Learn More
Ace Glass, Inc. 250ML 24/40-24/40 TRAP 250ML 24/40-24/40 TRAP
SDP
Catalog No. 50973373
Encompass_Preferred
250mL anti-climb rotary evaporator trap has upper tabulation to prevent migration of film and foam directly into the condenser. Trap has 24/40 standard taper joints top and bottom.
Catalog No. 50-973-373 Supplier Ace Glass, Inc. Supplier No. 670617
250mL anti-climb rotary evaporator trap has upper tabulation to prevent migration of film and foam directly into the condenser. Trap has 24/40 standard taper joints top and bottom.