Learn More
Ace Glass, Inc. Trap, bump, drain holes, 250mL, 24/40 top, 24/40 bottom
SDP
Catalog No. 509557382
Encompass_Preferred
250mL bump trap for use between a rotary evaporators steam tube and evaporator flask, preventing fluid from being drawn into the condenser. Trap has 24/40 top and bottom joints. Improved design incorporates two drain holes to expedite the return of solvent to the evaporator flask.
Catalog No. 50-955-7382 Supplier Ace Glass, Inc. Supplier No. 670514
250mL bump trap for use between a rotary evaporators steam tube and evaporator flask, preventing fluid from being drawn into the condenser. Trap has 24/40 top and bottom joints. Improved design incorporates two drain holes to expedite the return of solvent to the evaporator flask.