Learn More
Ace Glass, Inc. 250mL 24/40 addition funnel, PTFE stopcock plug, ungraduated
SDP
Catalog No. 50974728
Encompass_Preferred
250mL short stem addition funnel with 24/40 top and bottom joints, and a PTFE stopcock plug. Funnel is not graduated.
Catalog No. 50-974-728 Supplier Ace Glass, Inc. Supplier No. 725752
250mL short stem addition funnel with 24/40 top and bottom joints, and a PTFE stopcock plug. Funnel is not graduated.