Learn More
Chemglass Life Sciences 250mL Tube, Reaction, Airfree, Schlenk, 24/40 Outer Joint, 0-8mm Valve
SDP
Catalog No. AF0537A14
Encompass_Preferred
250mL Tube, Reaction, Airfree, Schlenk, 24/40 Outer Joint, 0-8mm Valve. Cylindrical shaped tube useful for storage or simple reactions with a standard taper outer joint at the top and a Chem-Vac™ Chem-Cap valve sidearm. Plug may be removed for sampling from tube while a purge of inert gas is maintained through the sidearm. Valve size is as listed.
Catalog No. AF0537A14 Supplier Chemglass Life Sciences Supplier No. AF0537A14
250mL Tube, Reaction, Airfree, Schlenk, 24/40 Outer Joint, 0-8mm Valve. Cylindrical shaped tube useful for storage or simple reactions with a standard taper outer joint at the top and a Chem-Vac™ Chem-Cap valve sidearm. Plug may be removed for sampling from tube while a purge of inert gas is maintained through the sidearm. Valve size is as listed.