Learn More
Ace Glass, Inc. 250mL 29/42-29/42 trap anti-climb
SDP
Catalog No. 50978864
Encompass_Preferred
250mL anti-climb rotary evaporator trap has upper tabulation to prevent migration of film and foam directly into the condenser. Trap has 29/42 standard taper joints top and bottom.
Catalog No. 50-978-864 Supplier Ace Glass, Inc. Supplier No. 670621
250mL anti-climb rotary evaporator trap has upper tabulation to prevent migration of film and foam directly into the condenser. Trap has 29/42 standard taper joints top and bottom.