Learn More
Chemglass Life Sciences 250mL 2-Neck Flask, Reaction, 14/20 Joint Size, Valve, Airfree, Schlenk
SDP
Catalog No. AF053022
Encompass_Preferred
250mL 2-Neck Flask, Reaction, 14/20 Joint Size, Valve, Airfree, Schlenk. Round bottom 2-neck flask having standard taper outer joints, both being of the same size. Available with a choice of a 2mm high vacuum glass stopcock having a Stainless Steel constant pressure retaining device or a 0-4mm Chem-Cap Airfree inlet valve. Side neck can be fitted with a CG-3022 septum stopper for sampling or cannula transfer.
Catalog No. AF053022 Supplier Chemglass Life Sciences Supplier No. AF053022
250mL 2-Neck Flask, Reaction, 14/20 Joint Size, Valve, Airfree, Schlenk. Round bottom 2-neck flask having standard taper outer joints, both being of the same size. Available with a choice of a 2mm high vacuum glass stopcock having a Stainless Steel constant pressure retaining device or a 0-4mm Chem-Cap Airfree inlet valve. Side neck can be fitted with a CG-3022 septum stopper for sampling or cannula transfer.