Learn More
Chemglass Life Sciences 250mL 3-Neck European Flask, 1-CN 14/20 Outer, 2-SN 14/20 Outer Angled 20°
SDP
Catalog No. CG157005
Encompass_Preferred
250mL 3-Neck European Flask, 1-CN 14/20 Outer, 2-SN 14/20 Outer Angled 20¿. European style flask with tapered walls and a shallow hemispherical bottom for more efficient mechanical or magnetic stirring. With standard taper outer joints on center and side necks.
Catalog No. CG157005 Supplier Chemglass Life Sciences Supplier No. CG157005
250mL 3-Neck European Flask, 1-CN 14/20 Outer, 2-SN 14/20 Outer Angled 20¿. European style flask with tapered walls and a shallow hemispherical bottom for more efficient mechanical or magnetic stirring. With standard taper outer joints on center and side necks.