Learn More
Ace Glass, Inc. 250ML 50MM BOTTLE 250ML 50MM BOTTLE
SDP
Catalog No. 50974957
Encompass_Preferred
250mL, 50mm air sampling collection bottle, usually connected to the bottom arm of a tee with reduced ends, to collect heavy particulates before they enter the manifold train.
Catalog No. 50-974-957 Supplier Ace Glass, Inc. Supplier No. 750115
250mL, 50mm air sampling collection bottle, usually connected to the bottom arm of a tee with reduced ends, to collect heavy particulates before they enter the manifold train.