Learn More
MAGIC BIOPROCESSING MAGic™Beads ACT 250mL
Catalog No. NC3724014
Encompass_Preferred
MAGic™ Beads ACT are super-paramagnetic porous beads designed for in-house coupling of ligands containing amino groups or thiols. Proteins, peptides, and other biomolecules can be covalently attached using a simple, sustainable process, all under physiological conditions and at room temperature, without hazardous reagents or organic solvents. Once coupled, the beads are ready for affinity purification or immunoprecipitation. Supplied as a 10% slurry, MAGic™ Beads ACT offer a versatile platform for developing your own magnetic bead-based applications.
Catalog No. NC3724014 Supplier MAGIC BIOPROCESSING Supplier No. 1325
MAGic™ Beads ACT are super-paramagnetic porous beads designed for in-house coupling of ligands containing amino groups or thiols. Proteins, peptides, and other biomolecules can be covalently attached using a simple, sustainable process, all under physiological conditions and at room temperature, without hazardous reagents or organic solvents. Once coupled, the beads are ready for affinity purification or immunoprecipitation. Supplied as a 10% slurry, MAGic™ Beads ACT offer a versatile platform for developing your own magnetic bead-based applications.