Learn More
Beckman Coulter Adapter, 392256
Catalog No. NC0730940
Encompass
The Beckman Coulter Microfuge 18 Centrifuge Adapter, catalog number 392256, is designed to adapt 0.2ml, 0.4ml, and 0.5ml microcentrifuge tubes to the Microfuge 18 centrifuge rotor. It allows centrifugation of up to 18 microcentrifuge tubes at one time. The adapter is made of durable polypropylene and is autoclavable. It fits securely into the Microfuge 18 rotor. This adapter provides a convenient way to spin standard microcentrifuge tubes in the Microfuge 18 benchtop centrifuge.
Catalog No. NC0730940 Supplier Beckman Coulter Supplier No. 392256
The Beckman Coulter Microfuge 18 Centrifuge Adapter, catalog number 392256, is designed to adapt 0.2ml, 0.4ml, and 0.5ml microcentrifuge tubes to the Microfuge 18 centrifuge rotor. It allows centrifugation of up to 18 microcentrifuge tubes at one time. The adapter is made of durable polypropylene and is autoclavable. It fits securely into the Microfuge 18 rotor. This adapter provides a convenient way to spin standard microcentrifuge tubes in the Microfuge 18 benchtop centrifuge.